Forskolin ≥98%
Bottle
Catalog # APOSBUP17787-200MG
New Product
Supplier: Apollo Scientific
Forskolin ≥98%
Catalog # APOSBUP17787-200MG
Supplier: Apollo Scientific
New Product
CAS Number: 66575-29-9
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
- CAS Number:66575-29-9
- Formula:C₂₂H₃₄O₇
- Molecular Weight:410,5 g/mol
Specifications
Specifications
- Pack type:Bottle



