L-Proline ≥98%
Bottle
Catalog # APOSBUP18747-500MG
New Product
Supplier: Apollo Scientific
L-Proline ≥98%
Catalog # APOSBUP18747-500MG
Supplier: Apollo Scientific
New Product
CAS Number: 147-85-3
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
- CAS Number:147-85-3
- Formula:C₅H₉NO₂
- Molecular Weight:115,13 g/mol
Specifications
Specifications
- Pack type:Bottle



