Dulcite ≥98%
Bottle
N° de catalogue APOSBUP16557-100MG
New Product
Fournisseur: Apollo Scientific
Dulcite ≥98%
N° de catalogue APOSBUP16557-100MG
Fournisseur: Apollo Scientific
New Product
Numéro CAS: 608-66-2
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:608-66-2
- Formula:C₆H₁₄O₆
- Molecular Weight:182,17 g/mol
Specifications
Specifications
- Pack type:Bottle



