L(+)-Ascorbic acid 99% USP, BP, Ph. Eur., pure, Pharma Grade
New Product
Fournisseur: Apollo Scientific
L(+)-Ascorbic acid 99% USP, BP, Ph. Eur., pure, Pharma Grade
Fournisseur: Apollo Scientific
New Product
Certificates
Jump to:
About this item
Usage: Biochemical/life science applications.
- Store at room temperature
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176,12412 g/mol
4 Options Available Below
Product Details & Documents
Usage: Biochemical/life science applications.
- Store at room temperature
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176,12412 g/mol
Product Family Options
Product Information
- Pack typeDisponibilitéPrix
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



