Order Entry
Belgium
Orders LinkContactUsLinkComponent
Phenolphthalein ≥98%

Phenolphthalein ≥98%

New Product
Supplier:   Apollo Scientific
Phenolphthalein ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Phenolphthalein ≥98%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
  • CAS Number:77-09-8
  • Formula:C₂₀H₁₄O₄
  • Molecular Weight:318,32 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 53,60 €
    53,60 €
    1 * 500 mg
       53,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 63,70 €
    63,70 €
    1 * 1 g
       63,70 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.