DBCO-PEG4-NHS ester
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/ life science applications.
- Store at −20 °C
Synonyms: Dibenzocyclooctyne-PEG4-succinimidyl Ester, 2,5-Dioxo-1-pyrrolidinyl 20-(11,12-didehydrodibenz[b,f]azocin-5(6H)-yl)-17,20-dioxo-4,7,10,13-tetraoxa-16-azaeicosanoate, DBCO-PEG4-succinimidyl Ester, ADIBO-PEG4-NHS, DIBACA-PEG4-NHS
- CAS Number:1427004-19-0
- Formula:C₃₄H₃₉N₃O₁₀
- MDL Number:MFCD26793797
- Molecular Weight:649,68756 g/mol
- Storage Temperature:Freezer
2 Options Available Below
Product Details & Documents
Usage: Biochemical/ life science applications.
- Store at −20 °C
Synonyms: Dibenzocyclooctyne-PEG4-succinimidyl Ester, 2,5-Dioxo-1-pyrrolidinyl 20-(11,12-didehydrodibenz[b,f]azocin-5(6H)-yl)-17,20-dioxo-4,7,10,13-tetraoxa-16-azaeicosanoate, DBCO-PEG4-succinimidyl Ester, ADIBO-PEG4-NHS, DIBACA-PEG4-NHS
- CAS Number:1427004-19-0
- Formula:C₃₄H₃₉N₃O₁₀
- MDL Number:MFCD26793797
- Molecular Weight:649,68756 g/mol
- Storage Temperature:Freezer



