Phenolphthalein TS, U.S.P., Test Solution (TS)
Catalog # SPCMP-127-500ML
New Product
Supplier: Spectrum Chemical
Phenolphthalein TS, U.S.P., Test Solution (TS)
Catalog # SPCMP-127-500ML
Supplier: Spectrum Chemical
New Product
CAS Number: 77-09-8
Jump to:
Product Details & Documents
Phenolphthalein TS, U.S.P. Test Solution is a ready-for-use solution for the purpose of conducting USP, National Formulary and assay tests and is prepared according to the specifications of the USP pharmacopeial convention.
Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
- CAS Number:77-09-8
- Formula:C₂₀H₁₄O₄
- MDL Number:MFCD00005913
- Molecular Weight:318,33 g/mol
- UN:UN1170



