Labscope 4.6 Software for Windows
Labscope 4.6 software for Windows, used to control and acquire images through connected and compatible microscopes, Compatible with Primo Star, Primovent, Axio Vert.A1, Axio Vert.A1 FL, Axio Vert.A1 MAT, Axio Vert.A1 FL-LED, Primotech, Stemi 305, Stemi 508, Axioscope 5 MAT, Axioscope 5 Pol, Axioscope vario, Stemi 305 cam, Axiolab 5, Axiolab 5 MAT, Axiolab 5 Pol, Axioscope 5, Primostar 3, Primostar 1, Axiovert 5, Axiovert 5 digital, Axiovert 7, LSM 990, Stemi 355
Catalog # ZEIS4101350052460
New Product
Supplier: ZEISS
Jump to:
Product Details & Documents
This combined system is used for in-vitro examination of images to obtain information about physiological or pathological processes or conditions.
Labscope for windows is intended for the use of professional users, among others in the field of life science, cytology and pathology.
The connected microscopes are themselves CE-IVD products. For IVD examinations, ensure that the displayed image on the monitor shows the equivalent information as visible through the eyepieces of the microscope. Labscope AI modules are not intended for IVD examinations and may only be used in research.
Specifications
- Description:Labscope 4.6 software for Windows, used to control and acquire images through connected and compatible microscopes
- For:Compatible with Primo Star, Primovent, Axio Vert.A1, Axio Vert.A1 FL, Axio Vert.A1 MAT, Axio Vert.A1 FL-LED, Primotech, Stemi 305, Stemi 508, Axioscope 5 MAT, Axioscope 5 Pol, Axioscope vario, Stemi 305 cam, Axiolab 5, Axiolab 5 MAT, Axiolab 5 Pol, Axioscope 5, Primostar 3, Primostar 1, Axiovert 5, Axiovert 5 digital, Axiovert 7, LSM 990, Stemi 355