Orders LinkContactUsLinkComponent
Optical Microscope
Optical Microscope
optical, LED illumination
Catalog # IDCPAF7115MZTW
New Product
Optical Microscope
Catalog # IDCPAF7115MZTW
New Product
Optical Microscope
optical, LED illumination
Price
814,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
The 5MP Edge AM7115MZTW - Edge provides macro-level imaging with zoomin capabilities. This microscope includes a polariser to reduce reflections and features FLC for adjustable illumination. AF7115MZTW is compatible with the optional WF-20 Wi-Fi streamer.
  • 5 Megapixel sensor
    Macro zoom optics
    Adjustable polariser
    Robust metal housing
    Flexible LED control (FLC)
    High optical resolution
    Adaptable interface
Spécifications
  • Magnification:
    Min.: 10 unit, Max.: 50 unit
  • Illumination:
    LED illumination
  • Type:
    optical
  • Cat. No.:
    IDCPAF7115MZTW
  • Observation method:
    Polarisation
  • Camera:
    Digital camera
  • Weight:
    97 g