Soder-Wick® Desoldering Braids
Product Details & Documents
Soder-Wick Rosin has the fastest wicking action, but rosin flux leaves behind residues that should be removed with a flux remover. Flux-Off Rosin remover is an ideal choice for your cleaning process.
Non corrosive ultra high purity Type R rosin flux
Minimises the risk of heat damage to the board
Will not leave ionic contamination on the boards
Soder-Wick brand desoldering wick offers the state of the art in desoldering technology. Soder-Wick is designed for today’s heat sensitive electronic components using lighter mass, pure copper braid construction that allows for better thermal conductivity, even at low temperatures. Soder-Wick responds faster than conventional desoldering braids thereby minimising overheating and preventing PCB damage.
Specifications:
MIL-F-14256 F type R flux
NASA-STD-8739.3 Soldered Electrical Connections
DOD-STD-883E, Method 2022
ANSI/IPC J STD-004, Type ROL0
Packaging: Each 25' or longer spool is individually packaged in a sealed bag. 5' and 10' bobbins have 2 packaging options.
Specifications
- Description:ESD-safe bobbins #5 brown, best for terminals; 10 bobbins per resealable pouch, 6 pouches; width: 3,7 mm; length: 1,5 m
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.