Order Entry
Belgium
Orders LinkContactUsLinkComponent
Panel Printer, 6290

Panel Printer, 6290

Panel printer
Catalog # SMEGWD-PRINT6290
New Product
Supplier:   SMEG
Panel Printer, 6290
Catalog # SMEGWD-PRINT6290
Supplier:  SMEG
New Product
Panel Printer, 6290

Panel printer

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Dot matrix thermal printing.

  • Integrated real-time clock

    Designed for panel fastening

Specifications
  • Description:
    Panel printer
  • Cat. No.:
    SMEGWD-PRINT6290
  • Ø:
    Roll: 50 mm
  • Dimensions:
    120L×170D×40H mm
  • Width:
    Roll: 57 mm