Order Entry
Belgium
Orders LinkContactUsLinkComponent
Panel Printer, 4290

Panel Printer, 4290

Panel printer
Catalog # SMEGWD-PRINT4290
New Product
Supplier:   SMEG
Panel Printer, 4290
Catalog # SMEGWD-PRINT4290
Supplier:  SMEG
New Product
Panel Printer, 4290

Panel printer

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Dot matrix thermal printing.

  • Integrated real-time clock

    Designed for panel fastening

     

Specifications
  • Description:
    Panel printer
  • Cat. No.:
    SMEGWD-PRINT4290
  • Ø:
    Roll: 50 mm
  • Dimensions:
    120L×170D×40H mm
  • Width:
    Roll: 57 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.