Orders LinkContactUsLinkComponent
Mobile Unit for Modules, E 941
Mobile Unit for Modules, E 941
Mobile unit for orthopedic instruments, stainless steel, PG 8527, PG 8528, PLW 8615, PLW 8616, PLW 8617
Catalog # MIEL12895230
New Product
Supplier:   MIELE
Mobile Unit for Modules, E 941
Catalog # MIEL12895230
Supplier:  MIELE
New Product
Mobile Unit for Modules, E 941
Mobile Unit for Modules, E 941
Mobile unit for orthopedic instruments, stainless steel, PG 8527, PG 8528, PLW 8615, PLW 8616, PLW 8617
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Mobile unit (E 941) for modules on 2 levels.
  • Suitable for up to 2 modules per level depending on the size of the load
    Coupling of the mobile unit to the water and drying air supply
    Large-capacity washer-disinfectors
    Reprocessing of laboratory glassware
    Magnetic strip for automatic mobile unit recognition (incl. magnets): 1
Specifications
  • Description:
    Mobile unit for orthopedic instruments, stainless steel
  • For:
    PG 8527, PG 8528, PLW 8615, PLW 8616, PLW 8617
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.