Order Entry
Belgium
Orders LinkContactUsLinkComponent
Low pH Phosphate SDS Sample Buffer

Low pH Phosphate SDS Sample Buffer

Low pH SDS phosphate sample buffer, 140 ml, 6,5
N° de catalogue EKSIC57805
New Product
Fournisseur:   EKSIGENT
Low pH Phosphate SDS Sample Buffer
N° de catalogue EKSIC57805
Fournisseur:  EKSIGENT
New Product
Numéro CAS:  
Low pH Phosphate SDS Sample Buffer

Low pH SDS phosphate sample buffer, 140 ml, 6,5

Numéro CAS:  
Prix
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

SCIEX Low pH SDS phosphate sample buffer, 140 ml (40 mM Phosphate, pH 6,5, 1% SDS) allows you to use SCIEX’s Gold Standard CE-SDS with a China Pharmacopeia compliant buffer.

Use of this buffer can increase non-reduced purity by decreasing method induced fragmentation. Less variability and lower RSDs means less troubleshooting and faster method development.

Specifications

Specifications

  • Description:
    Low pH SDS phosphate sample buffer, 140 ml
  • pH value:
    6,5

Specifications