Orders LinkContactUsLinkComponent
Handheld Digital Microscope Edge Plus AM8917MT4A
Handheld Digital Microscope Edge Plus AM8917MT4A
Handheld digital microscope, 33H×32D mm, AM8917MT4A, 8.0 Megapixel (3840 x 2160) on DinoCapture (Windows); 6.0 Megapixel (2880 x 2160) on DinoXcope (macOS)
N° de catalogue IDCPAM8917MT4A
New Product
Fournisseur:   Dino-Lite / IDCP B.V.
Handheld Digital Microscope Edge Plus AM8917MT4A
N° de catalogue IDCPAM8917MT4A
Fournisseur:  Dino-Lite / IDCP B.V.
New Product
Handheld Digital Microscope Edge Plus AM8917MT4A
Handheld digital microscope, 33H×32D mm, AM8917MT4A, 8.0 Megapixel (3840 x 2160) on DinoCapture (Windows); 6.0 Megapixel (2880 x 2160) on DinoXcope (macOS)
Prix
1 460,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The AM8917MT4A is a feature-packed microscope integrated with coaxial light and EDOF, answering needs for inspecting reflective surfaces and capturing all-in-focus images under 400x magnification. AM8917MT4A includes advanced features such as DPQ, EDR, AMR, and both auto and manual refocus.
High Quality Optics: The optics adopted in the Edge series reveals the finest details, answering the needs of the most demanding microscopy applications.
8.0 Megapixel: The 8.0-megapixel sensor captures a large field of view with a 16:9 aspect ratio and delivers natural-looking colors and enhanced image contrast at a resolution of up to 3840×2160.
Coaxial-Brightfield Illumination: The coaxial illumination uses a through-the-lens design, offering a compact yet high-quality solution for revealing imperfections and fine details on reflective surfaces.
Enhanced Flexible LED Control (eFLC, with coaxial light): Enhancing FLC capabilities, the eFLC can adjust the light intensity on two assigned groups of LED quadrants and the coaxial light, with 32 and 6 levels of adjustment respectively.
Extended Depth of Field (EDOF): Viewing rough surfaces with height ranges out of depth of focus, the EDOF can take several images at different focuses and stack them automatically within a click.
Extended Dynamic Range (EDR): Observing high contrast or reflective surfaces, the EDR can help to reveal the details of dark or bright areas by stacking images taken at different exposure levels.
​Auto and Manual Refocus: Using mouse inputs, the refocusing capability helps fine-tune the focus, either automatically by clicking at the point of interest or manually by scrolling the mouse.
​Automatic Magnification Readout (AMR): The Automatic Magnification Readout (AMR) detects the magnification in use, eliminating manual input errors and enhancing measurement accuracy in tasks with frequent magnification adjustments.
​Robust Metal Housing: The anodized aluminum alloy housing offers durability and EMI shielding, effectively guarding against impacts, abrasions, and electromagnetic noise.
Specifications
  • Magnification:
    415 ~ 460x
  • Model:
    AM8917MT4A
  • Resolution:
    8.0 Megapixel (3840 x 2160) on DinoCapture (Windows); 6.0 Megapixel (2880 x 2160) on DinoXcope (macOS)
  • Description:
    Handheld digital microscope, 33H×32D mm
  • Computer requirements:
    CPU: 2.9 GHz dual-core Intel Core i5 equivalent or greater; RAM: 4 GB or more; HD: 150 MB available space or more
  • Cat. No.:
    IDCPAM8917MT4A
  • Frame rate:
    10 fps at 8.0 MP / 6.0 MP, MJPEG; 25 fps at 2.0 MP, MJPEG