Orders LinkContactUsLinkComponent
Blocks for Vials
Blocks for Vials
New Product
Supplier:   VELP SCIENTIFIC
Blocks for Vials
Supplier:  VELP SCIENTIFIC
New Product
Certificates
Blocks for Vials
Blocks for Vials
9 Options Available Below
Product Details & Documents
These blocks are suitable for reaction vials, often used in chemistry and pharmaceutical applications for compound synthesis, extraction, or sample preparation.
Product Family Options
Product Information
  • Well size
    For
    No. of Holes
    Depth
    Availability
    Price
  • VELPA00000514
    VLS-W20-D120-A block
    VLS-W20-D120-A block
    Ø 12,7 mm
    12 mm vials
    20
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
    Specifications:
    New Product
    New Product
  • VELPA00000515
    VLS-W20-D150-A block
    VLS-W20-D150-A block
    Ø 15,8 mm
    15 mm vials
    20
    35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
    Specifications:
    New Product
    New Product
  • VELPA00000516
    VLS-W15-D160-A block
    VLS-W15-D160-A block
    Ø 16,4 mm
    16 mm vials
    15
    45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
    Specifications:
    New Product
    New Product
  • VELPA00000517
    VLS-W12-D170-A block
    VLS-W12-D170-A block
    Ø 17,8 mm
    17 mm vials
    12
    45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
    Specifications:
    New Product
    New Product
  • VELPA00000518
    VLS-W12-D190-A block
    VLS-W12-D190-A block
    Ø 19,7 mm
    19 mm vials
    12
    45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
    Specifications:
    New Product
    New Product
  • VELPA00000519
    VLS-W09-D210-A block
    VLS-W09-D210-A block
    Ø 21,7 mm
    21 mm vials
    9
    45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
    Specifications:
    New Product
    New Product
  • VELPA00000520
    VLS-W08-D230-A block
    VLS-W08-D230-A block
    Ø 23,8 mm
    23 mm vials
    8
    45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
    Specifications:
    New Product
    New Product
  • VELPA00000521
    VLS-W08-D250-A block
    VLS-W08-D250-A block
    Ø 25,8 mm
    25 mm vials
    8
    45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
    Specifications:
    New Product
    New Product
  • VELPA00000522
    VLS-W06-D280-A block
    VLS-W06-D280-A block
    Ø 28,8 mm
    28 mm vials
    6
    45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
    Specifications:
    New Product
    New Product