4 Point Comfort Plus Harness
Product Details & Documents
The 4 point comfort plus harness has been engineered from lightweight aluminium, optimising strength and durability.
Fully adjustable shoulder, chest and leg straps
Padded legs, lower back and shoulder straps for added comfort
Ergonomic quick release buckle
Inspection cover for protection of product label
Durable webbing with high breaking strength
Max rated load 100 kg
10 year shelf life from manufacturing date outlined in product label
UKCA marked
Retail box which aids presentation for retail sales
It provides four points of attachment, one rear D ring, two side D rings and an upper and lower chest D ring. Quick release buckles for easy fitting. Ideal for work positioning or suspension work.
Height safety is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas. Portwest offer a full range of height safety products for working at height.
Certifications: EN 813:2008; EN 1497:2007; EN 361:2002; EN 358:2018.
Specifications
- Attachment Points:4-point
- Description:Comfort plus harness, red
- Size:One size
- Weight:2,2 kg



