Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Masterflex® L/S® Variable Speed Analog Console Pump Systems

New Product
Masterflex® L/S® Variable Speed Analog Console Pump Systems
New Product
Certificates
Masterflex® L/S® Variable Speed Analog Console Pump Systems

5 Options Available Below

Product Details & Documents

Don’t lose control of your metering applications - digital display gives you precise control.

  • ABS plastic and aluminum housing
  • Accurate and repeatable settings with three-digit LED display of rpm

  • Pump fluid in either direction with reversible motor - perfect for purging tubing
  • Choose multichannel systems for synchronous flow from all channels
  • Maximise processes in a minimal footprint by stacking splash-resistant, IP 33-rated drives
  • Two ways to control speed, direction, and start/stop - use either the membrane keypad or remote-control capabilities

These L/S® precision variable-speed console drives offer precise ±0,25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.

Specifications for Product Family

Specifications

  • Model:
  • 77917-10
  • 77917-30
  • 77926-20
  • 77917-20
  • 77917-00
  • No. of channels:
  • 1
  • 1
  • 4
  • 1
  • 1
  • No. of rollers:
  • 3
  • 3
  • 6
  • 3
  • 3
  • Control type:
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Housing material:
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • Min. flow rate per channel:
  • 17 ml/min
  • 17 ml/min
  • 0,47 ml/min
  • 4,8 ml/min
  • 4,8 ml/min
  • Max. flow rate per channel:
  • 1700 ml/min
  • 1700 ml/min
  • 47 ml/min
  • 480 ml/min
  • 480 ml/min
  • Min. flow rate:
  • 17 ml/min
  • 0,47 ml/min
  • 4,8 ml/min
  • Drive IP rating:
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • Accuracy:
  • ±0,25% (1 rpm at 600 and 300 rpm; 0,1 rpm at 100 rpm)
  • ±0,25% (1 rpm at 600 and 300 rpm; 0,1 rpm at 100 rpm)
  • ±0,25% (1 rpm at 600 and 300 rpm; 0,1 rpm at 100 rpm)
  • ±0,25% (1 rpm at 600 and 300 rpm; 0,1 rpm at 100 rpm)
  • ±0,25% (1 rpm at 600 and 300 rpm; 0,1 rpm at 100 rpm)
  • Anti-drip:
  • No
  • No
  • No
  • No
  • No
  • Cloud connected:
  • No
  • No
  • No
  • No
  • No
  • Digital dispensing:
  • No
  • No
  • No
  • No
  • No
  • Drive:
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • Duty cycle:
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Min. RPM:
  • 6 rpm
  • 6 rpm
  • 1 rpm
  • 6 rpm
  • 6 rpm
  • Open-head sensor:
  • No
  • Yes
  • No
  • Yes
  • No
  • Power:
  • 90 - 260 VAC; 50/60 Hz; 1,8 A @ 115 V; 1,2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1,8 A @ 115 V; 1,2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1,8 A @ 115 V; 1,2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1,8 A @ 115 V; 1,2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1,8 A @ 115 V; 1,2 A @ 230 V
  • Pump series:
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • Remote I/O:
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • Reversible motor:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Tubing:
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16 tube set, pk of 8
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 16, 10'
  • Tubing sizes accepted:
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® two-stop tube sets: L/S® 13, 14, 16
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 13, 14, 16, 25, 17, 18
  • Washdown capable:
  • No
  • No
  • No
  • No
  • No
  • Current:
  • 1,8 A @ 115 V; 1,2 A @ 230 V
  • 1,8 A @ 115 V; 1,2 A @ 230 V
  • 1,8 A @ 115 V; 1,2 A @ 230 V
  • 1,8 A @ 115 V; 1,2 A @ 230 V
  • 1,8 A @ 115 V; 1,2 A @ 230 V
  • Frequency:
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • Voltage:
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years

Specifications

Product Family Options

Product Information

  • Maximum Flow Rate
    Pump head
    Height
    Length
    Availability
    Price
  • MFLX77917-10-EU
    L/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    L/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    1700 ml/min
    77200-62 (L/S® Easy-Load® II pump head)
    215,9 mm (8,5")
    339,9 mm (13,38")
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 490,00 €
    3 490,00 €
    1 * 1 ST
       3 490,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    MFLX77917-10-EU
    Model
    77917-10
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0,25% (1 rpm at 600 and 300 rpm; 0,1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1,8 A @ 115 V; 1,2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1,8 A @ 115 V; 1,2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • MFLX77917-30-EU
    L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    1700 ml/min
    77200-32 (L/S® Easy-Load® II OHS pump head)
    215,9 mm (8,5")
    339,9 mm (13,38")
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 660,00 €
    3 660,00 €
    1 * 1 ST
       3 660,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    MFLX77917-30-EU
    Model
    77917-30
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    17 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0,25% (1 rpm at 600 and 300 rpm; 0,1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1,8 A @ 115 V; 1,2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1,8 A @ 115 V; 1,2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • MFLX77926-20-EU
    L/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VAC
    L/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VAC
    188 ml/min
    07535-04 (L/S® 4-channel pump head)
    215,9 mm (8,5")
    349 mm (13,74")
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 330,00 €
    4 330,00 €
    1 * 1 ST
       4 330,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    MFLX77926-20-EU
    Model
    77926-20
    No. of channels
    4
    No. of rollers
    6
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    0,47 ml/min
    Max. flow rate per channel
    47 ml/min
    Min. flow rate
    0,47 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0,25% (1 rpm at 600 and 300 rpm; 0,1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
    Duty cycle
    Continuous
    Min. RPM
    1 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1,8 A @ 115 V; 1,2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16 tube set, pk of 8
    Tubing sizes accepted
    L/S® two-stop tube sets: L/S® 13, 14, 16
    Washdown capable
    No
    Current
    1,8 A @ 115 V; 1,2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • MFLX77917-20-EU
    L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VAC
    L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VAC
    480 ml/min
    77200-30 (L/S® Easy-Load® II OHS pump head)
    215,9 mm (8,5")
    352,6 mm (13,88")
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 660,00 €
    3 660,00 €
    1 * 1 ST
       3 660,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    MFLX77917-20-EU
    Model
    77917-20
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4,8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    4,8 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0,25% (1 rpm at 600 and 300 rpm; 0,1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1,8 A @ 115 V; 1,2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1,8 A @ 115 V; 1,2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • MFLX77917-00-EU
    L/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VAC
    L/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VAC
    480 ml/min
    77200-60 (L/S® Easy-Load® II pump head)
    215,9 mm (8,5")
    352,6 mm (13,88")
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 490,00 €
    3 490,00 €
    1 * 1 ST
       3 490,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    MFLX77917-00-EU
    Model
    77917-00
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4,8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0,25% (1 rpm at 600 and 300 rpm; 0,1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1,8 A @ 115 V; 1,2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1,8 A @ 115 V; 1,2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.