Order Entry
Belgium
Orders LinkContactUsLinkComponent
Chlorophenol red

Chlorophenol red

Bottle
Catalog # GENORC-649
Supplier:   G-Biosciences
Chlorophenol red
Catalog # GENORC-649
Supplier:  G-Biosciences
CAS Number:  4430-20-0
Chlorophenol red

Bottle

CAS Number:  4430-20-0
Price
567,00 €
Log in for availability
1 * 50 g
Product Details & Documents

Chlorophenol red is an indicator dye that changes colour from yellow to violet in the pH range 4,8 to 6,7.

  • Molecular Biology Grade
  • Brown crystalline powder
Synonyms:  3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
  • CAS Number:4430-20-0
  • Formula:C₁₉H₁₂Cl₂O₅S
  • Molecular Weight:423,27 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.