Temperature Data Loggers
New Product
Supplier: RAYPA
Jump to:
About this item
AISI-316L stainless steel disk temperature recorder with connection base and software.
- User-friendly operation
- Data logger available in multiple sizes to suit different needs
- Delivers high accuracy and precision
Recommended for autoclave validation and for monitoring the internal temperature of containers.
6 Options Available Below
Product Details & Documents
AISI-316L stainless steel disk temperature recorder with connection base and software.
- User-friendly operation
- Data logger available in multiple sizes to suit different needs
- Delivers high accuracy and precision
Recommended for autoclave validation and for monitoring the internal temperature of containers.
Product Family Options
Product Information
- W×D×HAvailabilityPrice
- ESPIBDLDISK_CLStainless steel and plastic connection base BDLDISK_CL, 120 g, for communication between the data logger and the software installed on a PCStainless steel and plastic connection base BDLDISK_CL, 120 g, for communication between the data logger and the software installed on a PC60×95×175 mm (External)Log in for availability1 * 1 STSpecifications:New ProductNew Product
- ESPIDLDISK3618_CLAISI-316L stainless steel disk temperature recorder DLDISK3618_CL, 80 g, for accurate and precise temperature monitoring; External (Ø×H): 36×18 mmAISI-316L stainless steel disk temperature recorder DLDISK3618_CL, 80 g, for accurate and precise temperature monitoring; External (Ø×H): 36×18 mmLog in for availability1 * 1 STSpecifications:New ProductNew Product
- ESPIBDL-DISK3618_CAISI-316L stainless steel disk temperature recorder with connection base and software BDL-DISK3618_CL, 200 g, for autoclave validation and for monitoring the internal temperature of containersAISI-316L stainless steel disk temperature recorder with connection base and software BDL-DISK3618_CL, 200 g, for autoclave validation and for monitoring the internal temperature of containers60×95×175 mm (External)Log in for availability1 * 1 PceSpecifications:New ProductNew Product
- ESPIBDL-DISK3618_TAISI-316L stainless steel disk temperature recorder with connection base and software, BDL-DISK3618_TL, 200 g, for autoclave validation and for monitoring the internal temperature of containersAISI-316L stainless steel disk temperature recorder with connection base and software, BDL-DISK3618_TL, 200 g, for autoclave validation and for monitoring the internal temperature of containers60×95×175 mm (External)Log in for availability1 * 1 STSpecifications:New ProductNew Product
- ESPIBDLDISK_TLStainless steel and plastic connection base for Communication between the data logger and the software installed on a PC, model: BDLDISK_TL, Weight: 120 gStainless steel and plastic connection base for Communication between the data logger and the software installed on a PC, model: BDLDISK_TL, Weight: 120 g60×95×175 mmLog in for availability1 * 1 STSpecifications:New ProductNew Product
- ESPIDLDISK3618_TLAISI-316 L stainless steel disk temperature recorder for Accurate and precise temperature monitoring, Ø×H: 36×18 mm, Model: DLDISK3618_TL, Weight: 80 gAISI-316 L stainless steel disk temperature recorder for Accurate and precise temperature monitoring, Ø×H: 36×18 mm, Model: DLDISK3618_TL, Weight: 80 gLog in for availability1 * 1 STSpecifications:New ProductNew Product