MPC Coupling Caps
About this item
MPC connectors are multi-purpose, open-format connectors for use in a wide variety of critical fluid handling applications. This particular type of connector is perfect for smaller bag systems for aliquoted media or other products stored in bags.
Ergonomic thumb latch: Easy to operate - even with gloved hands
- USP Class VI materials: Meet biocompatibility
- Sterilisable by autoclave, EtO, e-beam, or gamma: Robust and economical for single-use applications
- Parting line-free hose barb: Eliminates potential leak path
- BSE/TSE free materials: Meet ADCF requirements
MPC connectors add ease of use and security to the most critical fluid handling applications. Choose from a full line of male and female MPC connectors and configurations, including pressure sealing caps and plugs, in sizes to fit ¹/₈", ¹/₄", and ³/₈" tubing. MPC couplings offer optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks.
10 Options Available Below
Product Details & Documents
MPC connectors are multi-purpose, open-format connectors for use in a wide variety of critical fluid handling applications. This particular type of connector is perfect for smaller bag systems for aliquoted media or other products stored in bags.
Ergonomic thumb latch: Easy to operate - even with gloved hands
- USP Class VI materials: Meet biocompatibility
- Sterilisable by autoclave, EtO, e-beam, or gamma: Robust and economical for single-use applications
- Parting line-free hose barb: Eliminates potential leak path
- BSE/TSE free materials: Meet ADCF requirements
MPC connectors add ease of use and security to the most critical fluid handling applications. Choose from a full line of male and female MPC connectors and configurations, including pressure sealing caps and plugs, in sizes to fit ¹/₈", ¹/₄", and ³/₈" tubing. MPC couplings offer optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks.
Specifications for Product Family
Specifications
- Catalog No:
- MFLX30003-86
- MFLX30003-87
- MFLX30003-88
- MFLX30003-89
- MFLX30003-90
- MFLX30003-91
- MFLX30003-92
- MFLX30003-93
- MFLX30003-00
- MFLX30003-01
- Max. operating pressure:
- Up to 60 psi, 4,1 bar
- Up to 60 psi, 4,1 bar
- Up to 60 psi, 4,1 bar
- Up to 60 psi, 4,1 bar
- Up to 60 psi, 4,1 bar
- Up to 60 psi, 4,1 bar
- Up to 60 psi, 4,1 bar
- Up to 60 psi, 4,1 bar
- Max. temperature:
- 121 °C (250 °F)
- 121 °C (250 °F)
- 149 °C (300 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 149 °C (300 °F)
- 149 °C (300 °F)
- Min. temperature:
- −40 °C (−40 °F)
- −40 °C (−40 °F)
- −40 °C (−40 °F)
- −40 °C (−40 °F)
- −40 °C (−40 °F)
- −40 °C (−40 °F)
- −40 °C (−40 °F)
- −40 °C (−40 °F)
- Sterilisation:
- Gamma, autoclave
- Gamma, autoclave
- Gamma, autoclave
- Gamma, autoclave
- Gamma, autoclave
- Gamma, autoclave
- Gamma, autoclave
- Gamma, autoclave
Specifications
Product Family Options
MPX
Product Information
- TypeBarb size/tubing int.Ø (port 1)Connection TypeFitting materialFlow patternAvailabilityPrice
- MFLX30003-86¹/₂" Hose barb non-valved coupling body PC, MPX17803B25; 25 Pk¹/₂" Hose barb non-valved coupling body PC, MPX17803B25; 25 PkOpen¹/₂"Quick-disconnect to barbPCStraightLog in for availability1 * 25 ST124,00 €Specifications:New Product
More
Specifications
Cat. No.MFLX30003-86Max. operating pressureUp to 60 psi, 4,1 barMax. temperature121 °C (250 °F)Min. temperature−40 °C (−40 °F)SterilisationGamma, autoclaveNew Product - MFLX30003-87³/₈" Hose barb non-valved coupling insert PC, MPX22603M; 250 Pk³/₈" Hose barb non-valved coupling insert PC, MPX22603M; 250 PkOpen³/₈"Quick-disconnect to barbPCStraightLog in for availability1 * 250 ST1 200,00 €Specifications:New Product
More
Specifications
Cat. No.MFLX30003-87Max. operating pressureUp to 60 psi, 4,1 barMax. temperature121 °C (250 °F)Min. temperature−40 °C (−40 °F)SterilisationGamma, autoclaveNew Product - MFLX30003-88MPX coupling plug PS, MPX30039MB25; 25 PkMPX coupling plug PS, MPX30039MB25; 25 Pk——Quick-disconnect plugPPSUStraightLog in for availability1 * 25 ST359,00 €Specifications:New Product
More
Specifications
Cat. No.MFLX30003-88Max. operating pressureUp to 60 psi, 4,1 barMax. temperature149 °C (300 °F)Min. temperature−40 °C (−40 °F)SterilisationGamma, autoclaveNew Product - MFLX30003-89MPX coupling plug PC, MPX32003B25; 25 PkMPX coupling plug PC, MPX32003B25; 25 Pk——Quick-disconnect plugPCStraightLog in for availability1 * 25 ST148,00 €Specifications:New Product
More
Specifications
Cat. No.MFLX30003-89Max. operating pressureUp to 60 psi, 4,1 barMax. temperature121 °C (250 °F)Min. temperature−40 °C (−40 °F)SterilisationGamma, autoclaveNew Product
MPXK
Product Information
- TypeBarb size/tubing int.Ø (port 1)Connection TypeFitting materialFlow patternAvailabilityPrice
- MFLX30003-90¹/₂" Hose barb non-valved coupling body with lock PC, MPXK17803B25; 25 Pk¹/₂" Hose barb non-valved coupling body with lock PC, MPXK17803B25; 25 PkOpen¹/₂"Quick-disconnect to barbPCStraightLog in for availability1 * 25 ST206,00 €Specifications:New Product
More
Specifications
Cat. No.MFLX30003-90Max. operating pressureUp to 60 psi, 4,1 barMax. temperature121 °C (250 °F)Min. temperature−40 °C (−40 °F)SterilisationGamma, autoclaveNew Product - MFLX30003-91MPX Coupling cap, locking PC, MPXK32003B25; 25 PkMPX Coupling cap, locking PC, MPXK32003B25; 25 Pk——Quick-disconnect capPCStraightLog in for availability1 * 25 ST225,00 €Specifications:New Product
More
Specifications
Cat. No.MFLX30003-91Max. operating pressureUp to 60 psi, 4,1 barMax. temperature121 °C (250 °F)Min. temperature−40 °C (−40 °F)SterilisationGamma, autoclaveNew Product - MFLX30003-92MPX Coupling cap, locking PS, MPXK32039; 250 PkMPX Coupling cap, locking PS, MPXK32039; 250 Pk——Quick-disconnect capPPSUStraightLog in for availability1 * 250 ST5 880,00 €Specifications:New Product
More
Specifications
Cat. No.MFLX30003-92Max. operating pressureUp to 60 psi, 4,1 barMax. temperature149 °C (300 °F)Min. temperature−40 °C (−40 °F)SterilisationGamma, autoclaveNew Product - MFLX30003-93MPX Coupling cap, locking PS, MPXK32039B25; 25 PkMPX Coupling cap, locking PS, MPXK32039B25; 25 Pk——Quick-disconnect capPPSUStraightLog in for availability1 * 25 ST522,00 €Specifications:New Product
More
Specifications
Cat. No.MFLX30003-93Max. operating pressureUp to 60 psi, 4,1 barMax. temperature149 °C (300 °F)Min. temperature−40 °C (−40 °F)SterilisationGamma, autoclaveNew Product
Silicone Replacement Seals
Product Information
- TypeBarb size/tubing int.Ø (port 1)Connection TypeFitting materialFlow patternAvailabilityPrice
- MFLX30003-00SaniQuik USP class VI, ADCF silicone seal, ¾", 100 pk, siliconeSaniQuik USP class VI, ADCF silicone seal, ¾", 100 pk, siliconeSiliconeLog in for availability1 * 50 ST192,00 €Specifications:New Product
More
Specifications
Cat. No.MFLX30003-00Max. operating pressureMax. temperatureMin. temperatureSterilisationNew Product - MFLX30003-01SaniQuik USP class VI, ADCF silicone seal, 1-¹/₂"; 50 PkSaniQuik USP class VI, ADCF silicone seal, 1-¹/₂"; 50 PkSiliconeLog in for availability1 * 50 ST290,00 €Specifications:New Product
More
Specifications
Cat. No.MFLX30003-01Max. operating pressureMax. temperatureMin. temperatureSterilisationNew Product
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.