Order Entry
Belgium
Orders LinkContactUsLinkComponent
VU0155041 ≥99% positive allosteric mGluR4 agonist

VU0155041 ≥99% positive allosteric mGluR4 agonist

Marketsource
Supplier:   Abcam
VU0155041 ≥99% positive allosteric mGluR4 agonist
Supplier:  Abcam
Marketsource
Certificates
VU0155041 ≥99% positive allosteric mGluR4 agonist

2 Options Available Below

Product Details & Documents

Positive allosteric selective modulator at mGluR4 (EC50 values are 798 and 693 nM at human and rat mGluR4 receptors, respectively). It has advantages over PHCCC as it is more potent. Shows limited blood-brain barrier permeability. Active in behavioural models of Parkinson's disease following icv injection. Available in water-soluble form - please see VU0155041 sodium salt.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  cis-2-[[(3,5-Dichlorophenyl)amino]carbonyl]cyclohexanecarboxylic acid, rel-(1R,2S)-2-((3,5-Dichlorophenyl)carbamoyl)cyclohexane-1-carboxylic acid
  • CAS Number:1093757-42-6
  • Molecular Weight:316,2 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% (1R,2S)-2-[(3,5-dichlorophenyl)carbamoyl]cyclohexane-1-carboxylic acid
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 66,70 €
    66,70 €
    1 * 10 mg
       66,70 €

    Specifications:
    Marketsource
    Marketsource
  • Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 175,00 €
    175,00 €
    1 * 50 mg
       175,00 €

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.