Finasteride ≥99% Type II 5alpha-reductase inhibitor
Product Details & Documents
Type II 5α-reductase inhibitor (IC₅₀ values are 11,3 and 313 nM at type II and type I respectively). Reduces prostate gland size. Suppresses conversion of testosterone to dihydrotestosterone. Active in vivo and orally active.
Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:98319-26-7
- Molecular Weight:372,5 g/mol
Specifications
Specification Test Results
- Constituents::100% Finasteride



