Orders LinkContactUsLinkComponent
Polysorbate 80 solution 0,1
Polysorbate 80 solution 0,1
Marketsource
Supplier:   Abcam
Polysorbate 80 solution 0,1
Supplier:  Abcam
Marketsource
Certificates
3 Options Available Below
Product Details & Documents
Tween 80 Detergent, 10% Solution, ab291111 (previously known as Tween® 80, MegaPure™ Detergent, 10% Solution, 2126) is a non-ionic detergent useful for the extraction of membrane-bound proteins as well as isolation of nuclei from ma mMalian cell lines.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
Specifications for Product Family
Specification Test Results
  • Constituents::
    10% Tween 80
Product Family Options
Product Information
  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - 97,40 €
    97,40 €
    1 * 50 mL
       97,40 €
    Specifications:
    Marketsource
    Marketsource
  • Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 154,00 €
    154,00 €
    1 * 100 mL
       154,00 €
    Specifications:
    Marketsource
    Marketsource
  • Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 682,00 €
    682,00 €
    1 * 500 mL
       682,00 €