Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bromophenol blue Tracking dye

Bromophenol blue Tracking dye

Catalog # ABCAAB146339-25
Supplier:   Abcam
Bromophenol blue Tracking dye
Catalog # ABCAAB146339-25
Supplier:  Abcam
CAS Number:  115-39-9
Bromophenol blue Tracking dye
CAS Number:  115-39-9
Price
76,90 €
Log in for availability
1 * 25 g
Product Details & Documents

pH indicator (pH 3,0 (yellow) - 4,6 (blue)). Tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis. Migrates at approximately the same rate as 300 to 500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Molecular Weight:669,96 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1?6-benzoxathiole-1,1-dione
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.