Order Entry
Belgium
Orders LinkContactUsLinkComponent
Erythromycin Macrolide antibiotic

Erythromycin Macrolide antibiotic

Marketsource
Supplier:   Abcam
Erythromycin Macrolide antibiotic
Supplier:  Abcam
Marketsource
Certificates

2 Options Available Below

Product Details & Documents

Macrolide antibiotic. Inhibits cytochrome p450 3A4 and P-glycoprotein. Binds the 50S ribosomal subunit and inhibits protein synthesis in Gram-positive and Gram-negative bacteria.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (-)-Erythromycin
  • Molecular Weight:733,9 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Erythromycin
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 61,50 €
    61,50 €
    1 * 1 g
       61,50 €

    Specifications:
    Marketsource
    Marketsource
  • Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 97,40 €
    97,40 €
    1 * 5 g
       97,40 €

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.