Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Uric acid ≥99% Xanthine oxidase product

Catalog # ABCAAB146243-10
Supplier:   Abcam
Uric acid ≥99% Xanthine oxidase product
Catalog # ABCAAB146243-10
Supplier:  Abcam
CAS Number:  69-93-2
Uric acid ≥99% Xanthine oxidase product
CAS Number:  69-93-2
Price
76,90 €
Log in for availability
1 * 10 g
Product Details & Documents

Xanthine oxidase product. Final oxidation product of purine catabolism in humans and primates. Acts as a potent peroxynitrite scavenger and antioxidant.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  1H-Purine-2,6,8(3H,7H,9H)-trione
  • CAS Number:69-93-2
  • Molecular Weight:168,11 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Uric acid