Cholesterol ≥98% water-permeable lipid
Product Details & Documents
Water-permeable lipid and major component of all biological membranes. Distributed in body tissues, especially the brain and spinal cord. Inhibits basal channel activity in the brain. Use as emulsifying agent and liposome preparations. Active in vivoand in vitro.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:57-88-5
- Molecular Weight:386,7 g/mol
Specifications
Specification Test Results
- Constituents::100% Cholesterol



