Order Entry
Belgium
Orders LinkContactUsLinkComponent
Methoxy-X04 ≥98% amyloid beta fluorescent marker

Methoxy-X04 ≥98% amyloid beta fluorescent marker

Supplier:   Abcam
Methoxy-X04 ≥98% amyloid beta fluorescent marker
Supplier:  Abcam
Certificates
Methoxy-X04 ≥98% amyloid beta fluorescent marker

2 Options Available Below

Product Details & Documents

Blood-brain barrier permeable amyloid β fluorescent marker. Blue fluorophore. Derivative of Congo Red and Chrysamine G. Selectively binds fibrillar β-sheet deposits with high affinity (Ki =26,8 nM). Detects plaques, tangles and cerebrovascular amyloid in vivo.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  4,4'-((1E,1'E)-(2-Methoxy-1,4-phenylene)bis(ethene-2,1-diyl))diphenol, 4,4'-[(2-methoxy-1,4-phenylene)di-(1E)-2,1-ethenediyl]bisphenol
  • CAS Number:863918-78-9
  • Molecular Weight:344,4 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Methoxy-X04
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 200,00 €
    200,00 €
    1 * 5 mg
       200,00 €

  • Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 754,00 €
    754,00 €
    1 * 25 mg
       754,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.