Order Entry
Belgium
Orders LinkContactUsLinkComponent
Curcumin ≥98% Lipoxygenase and COX inhibitor, high purity

Curcumin ≥98% Lipoxygenase and COX inhibitor, high purity

Catalog # ABCAAB141921-10
Supplier:   Abcam
Curcumin ≥98% Lipoxygenase and COX inhibitor, high purity
Catalog # ABCAAB141921-10
Supplier:  Abcam
CAS Number:  458-37-7
Curcumin ≥98% Lipoxygenase and COX inhibitor, high purity
CAS Number:  458-37-7
Price
154,00 €
Log in for availability
1 * 10 mg
Product Details & Documents

High purity flavonoid derivative. Potent antioxidant, anti-inflammatory, analgesic and anticancer agent. Lipooxygenase and cyclooxygenase inhibitor (IC₅₀ values are 8 and 52 μM, respectively). Shows diverse range of actions including free radical scavenging activity, cardio and neuroprotective effects. Blood brain barrier permeable. Active in vitroand in vivo.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
  • CAS Number:458-37-7
  • Molecular Weight:368,4 g/mol
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.