Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Curcumin Lipooxygenase inhibitor

Curcumin Lipooxygenase inhibitor

Supplier:   Abcam
Curcumin Lipooxygenase inhibitor
Supplier:  Abcam
Certificates

2 Options Available Below

Product Details & Documents

High purity flavonoid derivative. Potent antioxidant, anti-inflammatory, analgesic and anticancer agent. Lipooxygenase and cyclooxygenase inhibitor (IC₅₀ values are 8 and 52 μM, respectively). Shows diverse range of actions including free radical scavenging activity, cardio and neuroprotective effects. Blood brain barrier permeable. Active in vitroand in vivo.

Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
  • CAS Number:458-37-7
  • Molecular Weight:368,4 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Curcumin
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 71,80 €
    71,80 €
    1 * 50 mg
       71,80 €

  • Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 200,00 €
    200,00 €
    1 * 250 mg
       200,00 €

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.