Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium deoxycholate Ionic detergent

Sodium deoxycholate Ionic detergent

Catalog # ABCAAB146305-50
Supplier:   Abcam
Sodium deoxycholate Ionic detergent
Catalog # ABCAAB146305-50
Supplier:  Abcam
CAS Number:  302-95-4
Sodium deoxycholate Ionic detergent
CAS Number:  302-95-4
Price
76,90 €
Log in for availability
1 * 50 g
Product Details & Documents

Water-soluble ionic detergent useful for extraction of membrane receptors and other plasma membrane proteins. Reported to induce tyrosine phosphorylation of β-catenin and its subsequent translocation to the nucleus, promoting activation of β-catenin signaling (effective concentration 5 to 50 μM).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
  • CAS Number:302-95-4
  • Molecular Weight:414,56 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Sodium deoxycholate
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.