Order Entry
Belgium
Orders LinkContactUsLinkComponent
Neutral red stain for histology

Neutral red stain for histology

Catalog # ABCAAB146365-25
Marketsource
Supplier:   Abcam
Neutral red stain for histology
Catalog # ABCAAB146365-25
Supplier:  Abcam
Marketsource
CAS Number:  553-24-2
Neutral red stain for histology
CAS Number:  553-24-2
Price
76,90 €
Log in for availability
1 * 25 g
Product Details & Documents

Commonly used histological stain for lysosymes that stains lysosomes red. Once taken up by cells the compound can be used to ascertain cell viability. Also used as a pH indicator.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
  • CAS Number:553-24-2
  • Molecular Weight:288,78 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% N8,N8,3-trimethyl-2,8-phenazinediamine monohydrochloride