Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

RIPA Buffer, Abcam

Marketsource
 :   Abcam
RIPA Buffer, Abcam
 :  Abcam
Marketsource
 
RIPA Buffer, Abcam

 

 

A ready-to-use buffer useful for lysing cells and tissues for radioimmunoprecipitation assay.

  • Constituents: 1% Alcohol Ethoxylate, 0.877% Sodium chloride, 0.788% Tris HCl, 0.5% Sodium deoxycholate, 0.1% Sodium lauryl sulfate

 : All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

 

 

 

  • Catalog No:
  • ABCAAB288006-100
  • ABCAAB288006-500
  • Storage temperature:
  • Store at -20°C.
  • Store at -20°C.

 

Product Family Options

 

  • Description
     
     
  • RIPA Buffer
       
       
    1 * 100 mL
    1 * 100 mL - 113,00 €
    113,00 €
    1 * 100 mL
       113,00 €

     
    Marketsource
     
    Specifications
    Cat. No.
    ABCAAB288006-100
    Storage temperature
    Store at -20°C.
    Marketsource
  • RIPA Buffer
       
       
    1 * 500 mL
    1 * 500 mL - 436,00 €
    436,00 €
    1 * 500 mL
       436,00 €

     
    Marketsource
     
    Specifications
    Cat. No.
    ABCAAB288006-500
    Storage temperature
    Store at -20°C.
    Marketsource