Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Permeabilization Buffer (Flow Cytometry), Abcam

Permeabilization Buffer (Flow Cytometry)
N° de catalogue ABCAAB314681-100ML
New Product
Fournisseur:   Abcam
Permeabilization Buffer (Flow Cytometry), Abcam
N° de catalogue ABCAAB314681-100ML
Fournisseur:  Abcam
New Product
Numéro CAS:  
Permeabilization Buffer (Flow Cytometry), Abcam

Permeabilization Buffer (Flow Cytometry)

Numéro CAS:  
Prix
190,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Permeabilization Buffer (ab314681) is a ready to use permeabilization buffer for intracellular immunofluorescence staining for microscopy or flow cytometry.

  • pH: 7.4

 : All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Permeabilization Buffer (Flow Cytometry)
  • Cat. No.:
    ABCAAB314681-100ML
  • Storage temperature:
    Store at -20°C.

Specifications