Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Fixation Buffer (Flow Cytometry), Abcam

Fixation Buffer (Flow Cytometry)
Catalog # ABCAAB314680-100ML
New Product
Supplier:   Abcam
Fixation Buffer (Flow Cytometry), Abcam
Catalog # ABCAAB314680-100ML
Supplier:  Abcam
New Product
CAS Number:  
Fixation Buffer (Flow Cytometry), Abcam

Fixation Buffer (Flow Cytometry)

CAS Number:  
Price
190,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Fixation Buffer (ab314680) is a ready-to-use formaldehyde-based fixation buffer for immunofluorescence cell staining for microscopy or flow cytometry.

  • pH: 7.4 Constituents: 5% Formaldehyde

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Fixation Buffer (Flow Cytometry)
  • Cat. No.:
    ABCAAB314680-100ML
  • Storage temperature:
    Store at Ambient.

Specifications