Orders LinkContactUsLinkComponent
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Supplier:  G-Biosciences
Certificates
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
2 Options Available Below
Product Details & Documents
Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
  • 1 M Tris·HCl (pH 7,4)
  • 1,5 M sodium chloride
Used to neutralize membrane-bound samples and stabilize probe-target duplexes. The Tris buffer maintains physiological pH, while the high salt concentration supports nucleic acid duplex stability and reduces nonspecific binding.
Documents
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • R017
    Hybridization neutralizing solution (1 M Tris.HCl (pH 7,4), 1,5 M NaCl)
    Hybridization neutralizing solution (1 M Tris.HCl (pH 7,4), 1,5 M NaCl)
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 88,10 €
    88,10 €
    1 * 1 L
       88,10 €
  • R018
    Hybridization neutralizing solution (1 M Tris.HCl (pH 7,4), 1,5 M NaCl)
    Hybridization neutralizing solution (1 M Tris.HCl (pH 7,4), 1,5 M NaCl)
    1 gal.
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 169,00 €
    169,00 €
    1 * 4 L
       169,00 €