Orders LinkContactUsLinkComponent
Switch-Grid™ Test Tube Rack, Rack Only
Switch-Grid™ Test Tube Rack, Rack Only
Polyoxymethylene, 318×111×67 mm
Catalog # 212-0691
Switch-Grid™ Test Tube Rack, Rack Only
Catalog # 212-0691
Switch-Grid™ Test Tube Rack, Rack Only
Switch-Grid™ Test Tube Rack, Rack Only
Polyoxymethylene, 318×111×67 mm
Price
45,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
The innovative Switch-Grid™ rack utilises interchangeable grids for ultimate flexibility. This split-frame rack holds up to two different tube sizes at the same time and allows users to quickly switch grids to hold other sizes as needed.
  • 10 to 30 mm diameter test tubes fit in 5 coloured-coded grid inserts
  • Can be easily cleaned with common lab detergents, ethanol, or isopropyl alcohol
  • Three-tier design securely holds tubes in place while non-slip rubber feet prevent sliding on the lab bench
  • Rack available without grids for use with spare grids
This rack is the smart, adaptable solution for optimising test tube organisation and streamlining laboratory workflows. Constructed from chemical-resistant polyoxymethylene plastic, both the frame and grids are autoclavable at 121 °C (250 °F).
Specifications
  • Material:
    Polyoxymethylene
  • W×D×H:
    318×111×67 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.