Order Entry
Belgium
ContactUsLinkComponent
 
DOPlify™ WGA V2 Kits
Catalog # BIOO4321-0020
Supplier:  BIOO SCIENTIFIC
CAS Number:  
undefined
DOPlify™ WGA V2 Kits
Catalog # BIOO4321-0020
Supplier:  BIOO SCIENTIFIC
CAS Number:  

Specifications

  • Description:
    DOPlify™ WGA v2 kit
  • No. of reactions:
    50

Specifications

About this item

The DOPlify® WGA V2 kit performs fast whole genome amplification on single cells or limited template DNA samples, allowing for the cell chromosome copy number status to be determined.

  • NGS compatible advanced DOP-PCR Fast whole genome amplification with protocol time 3 hours Reduced contamination risk with only two tube openings required and three reagents to add Can be fully automated with the JANUS® G3 liquid handler No amplification of no template control 2,5 μg of DNA from one cell

The DOPlify® WGA V2 kit offers the advantage of full mitochondrial DNA coverage and the ability to add custom primers to specially amplify regions of interest. Products of the DOPlify® WGA V2 kit can be used for a wide range of downstream processes, workflows, and NGS applications. The technology captures exclusive know-how for the lysis and whole genome amplification, with reagents provided in an easy-to-use kit format and less than three hours protocol time.

Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.