ContactUsLinkComponent
Proteometer-CV Starter Kit Reagent (60)
Proteometer-CV Starter Kit Reagent (60)
  NOVIKIT-PCV-00101B
 :  NOVILYTIC
New Product
 :  
undefined
Proteometer-CV Starter Kit Reagent (60)
  NOVIKIT-PCV-00101B
 :  NOVILYTIC
New Product
 :  
  • Description:
    Proteometer-CV starter kit reagent (60)
Fluorescent reagent portion of the Proteometer-CV starter kit. This is sold individually due to shipping and storage conditions.
  • No sample preparation required - analysis directly from clarified fermentation broth without protein A purification
  • Speed and efficiency - CV analysis in under 25 minutes
  • Simplified workflow - all the components you need for seamless operation
  • Exclusive technology - the only solution that performs charge variant analysis directly in clarified fermentation broth
  • Store at −20 °C
Crimped, sealed, ready for use with the Proteometer-CV starter kit (NOVIKIT-PCV-00101A), which is purchased separately. This kit is reagent-only and pairs with our Proteometer-CV starter kit.
Reagent to be stored at −20 °C.
 : Please add starter Proteometer-CV kit seperately (NOVIKIT-PCV-001-01A).
 :
Includes 1× Proteometer-CV fluorescent reagent vial. Allows for analysis up to 60 samples.
 : Packaged separately from the Proteometer-CV full kit due to storage conditions.