Order Entry
Belgium
Orders LinkContactUsLinkComponent
Proteometer-L buffer packets

Proteometer-L buffer packets

Proteometer-L Buffer Packets (7X)
Catalog # NOVIBUF-PL-001
New Product
Supplier:   NOVILYTIC
Proteometer-L buffer packets
Catalog # NOVIBUF-PL-001
Supplier:  NOVILYTIC
New Product
CAS Number:  
Proteometer-L buffer packets
Proteometer-L buffer packets

Proteometer-L Buffer Packets (7X)

CAS Number:  
Price
440,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

7 sealed buffer packets specifically formulated for use with Proteometer-L kit.

  • Buffer packets available for reorder when the Proteometer-L reactor and/or Proteometer-L reagent are not required

Intended for use with the Proteometer-L kit.

Ordering information: Reorder individually as needed.

Packaging: Supplied in a custom box with instructions for use. Store at room temperature.

Specifications

Specifications

  • Description:
    Proteometer-L Buffer Packets (7X)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.