Proteometer-L Starter Kit Reagent (200)
Catalog # NOVIKIT-PL-002-01B
Supplier: NOVILYTIC
New Product
CAS Number:
Specifications
- Description:Proteometer-L starter kit reagent (200)
Specifications
About this item
Fluorescent reagent for the Proteometer-L Starter Kit. This is sold separately due to shipping conditions.
- One assay for two critical attributes - titer + aggregate measurement
- No protein A purification required: Reduces costs
- No sample pretreatment: Reduces hours of sample prep
- No method development: Reduces time until method usage
- Direct analysis from CFB: HCP removal not needed
- Plug-and-play for any LC with fluorescence detector: Easy adoption, no software needed
- Low sample volume: Reduces impact on processing
Crimped, sealed, ready for use with the Proteometer-L starter kit (NOVIKIT-PL-002-01A), which is purchased separately. This kit is reagent-only and pairs with our Proteometer-L starter kit.
Store at −20 °C.
Ordering information: Please add starter Proteometer-L kit seperately (NOVIKIT-PL-002-01A)
Delivery: Includes 4× Proteometer-L fluorescent reagent vials. Allows for analysis up to 200 samples.
Packaging: Packaged separately from the Proteometer-L starter kit due to storage conditions.