Order Entry
Belgium
Orders LinkContactUsLinkComponent
Brady® i75 Series R4400 Ribbon for the i7500 Label Printer

Brady® i75 Series R4400 Ribbon for the i7500 Label Printer

Window cleaning squeegee, 46 cm (18"), Stainless steel
Catalog # CONTWCSQ0018
New Product
Supplier:   CONTEC
Brady® i75 Series R4400 Ribbon for the i7500 Label Printer
Catalog # CONTWCSQ0018
Supplier:  CONTEC
New Product
Brady® i75 Series R4400 Ribbon for the i7500 Label Printer
Brady® i75 Series R4400 Ribbon for the i7500 Label Printer

Window cleaning squeegee, 46 cm (18"), Stainless steel

Price
330,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

This stainless steel squeegee is lightweight, easy to use, and compatible with QuickConnect™ handles. It features a replaceable rubber blade, a secure press-down clip, and a screw-free channel for fast blade changes. The ergonomic, non-slip grip provides comfort and control for efficient cleanroom cleaning.

  • Lightweight, stainless-steel tool with rubber grip

  • Rubber grip prevents tool from slipping when used with disinfectants
  • Maneuvers easily to use vertically and horizontally, allowing users to clean tall windows and hard to reach areas
  • All components of the squeegee are compatible with common cleanroom cleaners and disinfectants
  • Press down handle clip allows for easy installation, preventing the rubber blade from moving during use
  • Screw-free channel allows blades to be replaced quickly and easily
  • Squeegee can be autoclaved at 121° C for 30 minutes
  • Ideal for use in all areas of the cleanroom

The stainless steel squeegee is a durable, lightweight tool designed for efficient cleanroom cleaning and liquid removal. Its rubber grip ensures a secure hold, even when used with disinfectants, while the screw-free channel allows for quick blade changes. Compatible with Contec QuickConnect™ handles, it easily reaches high windows and tight spaces. Fully autoclavable at 121° C for 30 minutes, it meets strict cleanroom standards, making it ideal for use in pharmaceutical, biotech, and microelectronics environments.

Specifications
  • Description:
    Window cleaning squeegee
  • Length:
    46 cm (18")
  • Material:
    Stainless steel
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.