Order Entry
Belgium
Orders LinkContactUsLinkComponent
QuEChERS Dispersive Solid Phase Extraction (SPE) Kit

QuEChERS Dispersive Solid Phase Extraction (SPE) Kit

QuEChERS Dispersive SPE kit, 5 g sample, 50 ml
Catalog # HYPES1-15-CN-KIT
QuEChERS Dispersive Solid Phase Extraction (SPE) Kit
Catalog # HYPES1-15-CN-KIT
CAS Number:  
QuEChERS Dispersive Solid Phase Extraction (SPE) Kit

QuEChERS Dispersive SPE kit, 5 g sample, 50 ml

CAS Number:  
Price
217,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

The QuEChERS Extraction Kit is designed for the efficient extraction of pesticides from food samples, ensuring compliance with the 2025 China Pharmacopeia.

  • Efficiently extracts pesticides from food sample

    Ensures compliance with the 2025 China Pharmacopeia

    Prepacked for convenience and ease of use

    Provides reliable, high-quality results for analytical testing

Sorbent: Anhydrous magnesium sulphate, Primary/Secondary amine End capped C18, Graphitised carbon black

Tube size: 15 ml

Delivery information: Includes kit with 900 mg MgSO₄, 300 mg PSA, 300 mg C18, 100 mg Si, 4 mg GCB, prefilled in 15 ml QuEChERS tubes.

Packaging: Pack of 50.

Specifications

Specifications

  • Description:
    QuEChERS Dispersive SPE kit, 5 g sample, 50 ml

Specifications