ContactUsLinkComponent
Service for Temperature and Humidity Logger, EBI 20-TH1
Service for Temperature and Humidity Logger, EBI 20-TH1
Catalog # EBRO1030-3323
Supplier:  EBRO
Service for Temperature and Humidity Logger, EBI 20-TH1
Catalog # EBRO1030-3323
Supplier:  EBRO
Specifications
  • Description:
    Accredited calibration service for temperature and humidity loggers of the product series: EBI 20, EBI 300, EBI 25, EBI 310, consisting of DIN EN ISO/IEC 17025 (accredited calibration according to DAkkS specifications); Calibration at 3 temperature points and the calibration of the additional measurand