Orders LinkContactUsLinkComponent
Stackable Storage Boxes, Polyethylene
Stackable Storage Boxes, Polyethylene
New Product
Supplier:   MULTIROIR
Stackable Storage Boxes, Polyethylene
Supplier:  MULTIROIR
New Product
Certificates
Stackable Storage Boxes, Polyethylene
Stackable Storage Boxes, Polyethylene
3 Options Available Below
Product Details & Documents
White polyethylene bins designed for laboratory environments. Smooth walls and rounded edges for easy, hygienic cleaning. Sturdy, stackable and available in three sizes. Suitable for intensive professional use.
  • Three sizes available
  • Closed handles with rounded edges
  • Smooth walls and rounded inner corners for optimal cleaning
  • Stackable to save space
  • Compatible with the requirements of laboratories and sensitive industries
  • Variable operating temperature depending on model
  • Solid walls/base
Made from high-quality polyethylene, these solid-sided white containers are perfectly suited to laboratory environments requiring cleanliness, strength and practicality. Their smooth inner surface and rounded edges enable fast, hygienic cleaning without residue retention, essential for preserving the integrity of samples or products handled.
Thanks to their robust design, these bins resist abrasion and are suitable for intensive, repeated use. Stackable to optimise storage space, they are available in three sizes and feature ergonomic closed handles for safe handling, even when wearing gloves. Recommended for use in laboratories, food processing and industrial environments.
Product Family Options
Product Information
  • Colour
    W×D×H
    Material
    Weight
    Availability
    Price
  • 216-3258
    Stackable storage box, temp. of use: −10...+50 °C
    Stackable storage box, temp. of use: −10...+50 °C
    White
    668×452×403 mm (ext.)
    Polyethylene
    3690 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
    Specifications:
    New Product
    New Product
  • 216-3257
    Stackable storage box, temp. of use: −30...+30 °C
    Stackable storage box, temp. of use: −30...+30 °C
    White
    660×450×225 mm (ext.)
    Polyethylene
    2360 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,00 €
    63,00 €
    1 * 1 ST
       63,00 €
    Specifications:
    New Product
    New Product
  • 216-3256
    Stackable storage box, temp. of use: −30...+30 °C
    Stackable storage box, temp. of use: −30...+30 °C
    White
    668×445×122 mm (ext.)
    Polyethylene
    1900 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,40 €
    56,40 €
    1 * 1 ST
       56,40 €
    Specifications:
    New Product