ContactUsLinkComponent
Tork S1 Floor Protector for Tork Hand Sanitiser Dispenser Stand, Black
Tork S1 Floor Protector for Tork Hand Sanitiser Dispenser Stand, Black
Catalog # SCAA511049
Supplier:  Essity
New Product
undefined
Tork S1 Floor Protector for Tork Hand Sanitiser Dispenser Stand, Black
Catalog # SCAA511049
Supplier:  Essity
New Product
Specifications
  • Colour:
    Black
  • Description:
    Floor protector, compatible with Tork S1, S4, S2 refills
Specifications
About this item
Tork floor protector for Tork hand sanitiser dispenser stand black S1, discreet elevated design. It prevent hand sanitiser spills with this floor protector, which is designed for use with Tork hand sanitiser dispenser stands.
  • Enjoy a seamless look with your existing interior decor, thanks to its sleek, discreet design
  • Boost cleaning efficiency with this easy-to-wipe model
  • Reduce maintenance, due to the floor protector's robust construction
    Material: Metal/plastic
    Suitable for S1 refills from Tork
Prevent unsightly hand sanitiser spills and reduce the risk of slipping with the Tork floor protector. This efficient protector fits neatly onto any Tork hand sanitiser dispenser stand to keep floors mess-free and guests safe. Best of all, it has a discreet design that's barely noticeable when guests use the hand sanitiser dispenser. It's also sturdy to ensure long-lasting use while reducing everyday maintenance.