ContactUsLinkComponent
HPLC Columns, Reprospher 100 Biphenyl
HPLC Columns, Reprospher 100 Biphenyl
Supplier:  DR MAISCH
New Product
Certificates
HPLC Columns, Reprospher 100 Biphenyl
Supplier:  DR MAISCH
New Product
Specifications
  • Catalog No:
  • MAISRS15.BPE.S1520
  • MAISRS15.BPE.S1503
  • Endcapped:
  • Double
  • Double
  • pH range:
  • 2 - 8
  • 2 - 8
  • Surface area:
  • 350 m²/g
  • 350 m²/g