ContactUsLinkComponent
AlphaTec® 2000 213 Standard Apron Stitched
  ANSEWH20S-00213-00
 :  Ansell
New Product
AlphaTec® 2000 213 Standard Apron Stitched
  ANSEWH20S-00213-00
 :  Ansell
New Product
  • Clothing Size:
    Universal
  • Packed:
    200 pieces
  • Garment application:
  • Gender:
  • Colour:
    White
  • Description:
    AlphaTec® 2000 213 standard apron stiched
  • Environmentally Preferable:
Apron with protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards.
  • Low-linting microporous laminate: Breathable comfort with lower contamination risks
    Optimised fit: An ideal balance of comfort and safety
Specialised protection: This AlphaTec® apron ensures barrier protection and resists saturation when confronted with low-concentration liquid chemicals, liquid and particulate biological hazards.
Heightened breathability: The use of microporous laminate technology in its fabrication makes for a protective apron that enables water vapor (perspiration) to escape easily.
Reduced contamination risks: Due to the low-linting nature of this protective apron’s microporous polyethylene laminate nonwoven material, potential contamination risks are also reduced.
Optimised fit: It is also designed to offer wearers a perfect combination of fit, comfort and personal protection.
Recommended for
-Clean room cleaning and preparing
-Production line support & maintenance
-Maintenance of plant and machines
-Veterinary Services
-Cleaning of plant and machines
-Body painting including surface inspection
 :
EN ISO 13688:2013.