Order Entry
Belgium
ContactUsLinkComponent
Tork T4 Premium Soft Conventional Toilet Roll
Tork T4 Premium Soft Conventional Toilet Roll
Catalog # SCAA110777
Supplier:  Essity
New Product
undefined
Tork T4 Premium Soft Conventional Toilet Roll
Catalog # SCAA110777
Supplier:  Essity
New Product

Specifications

  • Description:
    Premium conventional toilet paper roll, refill, white, compatible with Tork T4 dispensers
  • L×W:
    66000×94 mm

Specifications

About this item

Offer guests the comfort of home away from home with Advanced Tork traditional toilet paper. This toilet paper strikes a good balance between cost and performance for low-traffic washrooms.

  • Affordable paper that offers good value for money

  • Premium decor embossing for a nice look and more bulky feel

    Soft tissue with high brightness for a lasting impression

  • 6 packs, 9 rolls each

    600 sheets each, 2 ply

    Premium quality

    Sheet length 110 mm

    Core inner diameter 40 mm

Certifications:

EU Ecolabel | SE/004/001. FSC MIX Credit | SA-COC-008266.